siRNA Details
| Gene Symbol: SLC25A22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191060.1 | NP_001177989.1 | 6 | 854 | ||
| NM_001191061.1 | NP_001177990.1 | 6 | 523 | |||
| NM_024698.5 | NP_078974.1 | 6 | 691 | |||
| XM_011520369.1 | XP_011518671.1 | 6 | 565 | |||
| XM_011520370.1 | XP_011518672.1 | 6 | 633 | |||
| XM_011520371.1 | XP_011518673.1 | 6 | 518 | |||
| GenBank mRNA | AJ428202.1 | - | 5 | 411 | ||
| AK023106.1 | - | 5 | 411 | |||
| AK290481.1 | - | 6 | 854 | |||
| BC019033.2 | - | 6 | 489 | 4577143 | ||
| BC023545.2 | - | 6 | 649 | 4110556 | ||
| BC024212.2 | - | 2 | 32 | |||

SLC25A22
Genomic Map