siRNA Details
| Gene Symbol: ZNF703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025069.2 | NP_079345.1 | 2 | 2160 | ||
| GenBank mRNA | AK024361.1 | - | 2 | 2128 | ||
| BC032534.1 | - | 2 | 2089 | 5527569 | ||
| BC084581.1 | - | 3 | 841 | |||
| BM770319.1 | - | 1 | 54 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001110508, NM_001101502.

ZNF703
Genomic Map