This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FUZ SIRNA Assays ›
Gene Symbol:
FUZ
Gene Name:
fuzzy planar cell polarity protein
Gene Aliases:
FY, NTD
Chromosome Location:
Chr.19: 49806866 - 49817376 on Build GRCh38
UniGene ID:
Hs.288800
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s37041

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FUZ
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171937.1 NP_001165408.1 2 370
NM_025129.4 NP_079405.2 2 370
NR_033269.1 2 370
XM_006723399.3 XP_006723462.1 2 348
XM_011527339.1 XP_011525641.1 2 348
XM_011527340.1 XP_011525642.1 2 210
XM_011527341.2 XP_011525643.1 2 274
XM_011527342.1 XP_011525644.1 2 329
XM_011527343.1 XP_011525645.1 2 348
XM_011527345.1 XP_011525647.1 2 329
XM_011527347.1 XP_011525649.1 3 287
XM_017027319.1 XP_016882808.1 2 210
XM_017027320.1 XP_016882809.1 2 348
XM_017027322.1 XP_016882811.1 2 210
XM_017027323.1 XP_016882812.1 2 348
GenBank mRNA AK026341.1 - 2 344
AK057964.1 - 2 250
AK295072.1 - 2 369
AK315445.1 - 2 348
BC004445.1 - 2 271 2819805
BC010092.2 - 2 235 3354417
BC016793.2 - 2 271 2819805

Back To Top

More Information


Gene Ontology Categories:

Function(s)

establishment of planar polarity
neural tube closure
hair follicle development
negative regulation of cell proliferation
regulation of smoothened signaling pathway
embryonic body morphogenesis
protein transport
negative regulation of cell migration
nonmotile primary cilium assembly
cilium assembly
positive regulation of cilium assembly
embryonic skeletal system morphogenesis
negative regulation of canonical Wnt signaling pathway
negative regulation of neural crest formation
negative regulation of fibroblast growth factor receptor signaling pathway involved in neural plate anterior/posterior pattern formation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline