This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PAAF1 SIRNA Assays ›
Gene Symbol:
PAAF1
Gene Name:
proteasomal ATPase associated factor 1
Gene Aliases:
PAAF, Rpn14, WDR71
Chromosome Location:
Chr.11: 73876699 - 73927736 on Build GRCh38
UniGene ID:
Hs.525017
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s37097

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PAAF1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267803.1 NP_001254732.1 12 1158
NM_001267804.1 NP_001254733.1 12 1149
NM_001267805.1 NP_001254734.1 12 1145
NM_001267806.1 NP_001254735.1 11 1032
NM_025155.2 NP_079431.1 11 1089
XM_005274308.2 XP_005274365.2 9 1681
XM_005274309.1 XP_005274366.1 10 948
XM_017018357.1 XP_016873846.1 12 1077
XM_017018358.1 XP_016873847.1 12 1082
XM_017018359.1 XP_016873848.1 9 1640
GenBank mRNA AF087895.1 - 12 1083
AK021910.1 - 11 1070
AK222501.1 - 11 1070
AK225520.1 - 11 1057
AK298258.1 - 11 1032
AK309133.1 - 9 1296
AK316394.1 - 11 1303
BC006142.2 - 11 1042 3937356
BC021541.1 - 11 1053 3942680
BC028628.1 - 12 1123 5263875

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

viral process
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-98cc9b549-jkfrb:80/100.66.76.221:80.
git-commit: 5e7f4b920385fdbbb5f8d978f1c7c2eb3f871195
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-Offline