siRNA Details
| Gene Symbol: UBXN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171091.1 | NP_001164562.1 | 6 | 577 | ||
| NM_025241.2 | NP_079517.1 | 6 | 679 | |||
| XM_017027325.1 | XP_016882814.1 | 7 | 1110 | |||
| GenBank mRNA | AF272893.1 | - | 6 | 678 | ||
| AF272894.1 | - | 6 | 568 | |||
| AK090473.1 | - | 1 | 2023 | |||
| AL833695.1 | - | 1 | 534 | |||
| BC007414.2 | - | 2 | 139 | |||
| BC008288.1 | - | 6 | 577 | 3659797 | ||

UBXN6
Genomic Map