This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ST6GALNAC5 SIRNA Assays ›
Gene Symbol:
ST6GALNAC5
Gene Name:
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
Gene Aliases:
SIAT7-E, SIAT7E, ST6GalNAcV
Chromosome Location:
Chr.1: 76867408 - 77069167 on Build GRCh38
UniGene ID:
Hs.149201
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s37829

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ST6GALNAC5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320273.1 NP_001307202.1 4 701
NM_001320274.1 NP_001307203.1 3 593
NM_030965.2 NP_112227.1 5 1111
GenBank mRNA AJ507292.1 - 5 855
AK056241.1 - 5 1078
AK095758.1 - 6 997
AK295019.1 - 6 954
AK300902.1 - 4 626
BC001201.1 - 5 1018 3356535
DA166913.1 - 3 552

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001271361, AJ646872.
This siRNA also targets the following transcripts in Mouse: NM_012028, BC055737, AK042683, AB030836, AK038434, and others.

Gene Ontology Categories:

Function(s) Process(es)

ganglioside biosynthetic process
glycosphingolipid biosynthetic process
oligosaccharide metabolic process
oligosaccharide biosynthetic process
protein N-linked glycosylation via asparagine
sialylation
alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase activity
sialyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-xwmhz:80/100.66.76.181:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0