This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMUB1 SIRNA Assays ›
Gene Symbol:
TMUB1
Gene Name:
transmembrane and ubiquitin like domain containing 1
Gene Aliases:
C7orf21, DULP, SB144
Chromosome Location:
Chr.7: 151081085 - 151083533 on Build GRCh38
UniGene ID:
Hs.726215
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s38014

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMUB1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136044.1 NP_001129516.1 3 555
NM_031434.3 NP_113622.1 3 1018
GenBank mRNA AK314035.1 - 3 783
AY037155.1 - 3 536
AY358481.1 - 3 514
AY603380.1 - 2 540
BC000936.1 - 3 555 3449979
BC033182.1 - 3 693 4586932

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ER-associated ubiquitin-dependent protein catabolic process
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-wcshf:80/100.66.76.50:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0