This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM103A1 SIRNA Assays ›
Gene Symbol:
FAM103A1
Gene Name:
family with sequence similarity 103 member A1
Gene Aliases:
C15orf18, HsT19360, RAM
Chromosome Location:
Chr.15: 82986203 - 82991057 on Build GRCh38
UniGene ID:
Hs.80624
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s38061

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM103A1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031452.3 NP_113640.1 3 279
GenBank mRNA AK027095.1 - 3 240
BC003627.2 - 1 239 2989772
BC027181.1 - 1 231 3924570
BC112329.1 - 1 248 5578103

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

7-methylguanosine mRNA capping
methylation
recruitment of mRNA capping enzyme to RNA polymerase II holoenzyme complex
RNA binding
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-gdxnh:80/100.66.76.251:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0