This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF484 SIRNA Assays ›
Gene Symbol:
ZNF484
Gene Name:
zinc finger protein 484
Gene Aliases:
BA526D8.4
Chromosome Location:
Chr.9: 92834262 - 92878060 on Build GRCh38
UniGene ID:
Hs.668378
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s38174

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF484
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007101.2 NP_001007102.1 4 697
NM_001261458.1 NP_001248387.1 4 767
NM_001261459.1 NP_001248388.1 6 838
NM_001261460.1 NP_001248389.1 6 768
NM_031486.2 NP_113674.1 5 742
XM_006717299.3 XP_006717362.1 5 763
XM_011519073.2 XP_011517375.1 6 879
XM_011519074.2 XP_011517376.1 5 652
XM_011519075.2 XP_011517377.1 6 808
XM_017015178.1 XP_016870667.1 7 834
XM_017015179.1 XP_016870668.1 7 682
XM_017015180.1 XP_016870669.1 5 612
XM_017015181.1 XP_016870670.1 7 846
GenBank mRNA AK091203.1 - 4 647
AK292997.1 - 7 826
AK299274.1 - 4 665
AK309617.1 - 5 742
AL833202.1 - 5 712
BC112394.1 - 4 592 40008083
BC112396.1 - 4 592 40008084

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-n2s6g:80/100.66.79.83:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0