This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RAB6C SIRNA Assays ›
Gene Symbol:
RAB6C
Gene Name:
RAB6C, member RAS oncogene family
Gene Aliases:
WTH3
Chromosome Location:
Chr.2: 129979662 - 129982738 on Build GRCh38
UniGene ID:
Hs.591552
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s38489

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RAB6C
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032144.2 NP_115520.2 1 1647
GenBank mRNA AK055504.1 - 2 1034
AL136727.1 - 1 1648
BC120999.1 - 1 1485 40116526

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

retrograde vesicle-mediated transport, Golgi to ER
intra-Golgi vesicle-mediated transport
small GTPase mediated signal transduction
regulation of centrosome duplication
cell cycle process
retrograde transport, endosome to Golgi
response to drug
GTPase activity
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline