siRNA Details
| Gene Symbol: DGCR6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033257.3 | NP_150282.2 | 1 | 183 | ||
| GenBank mRNA | AF228708.1 | - | 1 | 163 | ||
| AK001528.1 | - | 1 | 124 | |||
| AK289952.1 | - | 1 | 101 | |||
| AK307210.1 | - | 1 | 119 | |||
| AK310899.1 | - | 1 | 158 | |||
| BC000682.2 | - | 1 | 119 | 3349309 | ||
| CR456361.1 | - | 1 | 88 | |||
| KJ535014.1 | - | 1 | 66 | |||

DGCR6L
Genomic Map