This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TSLP SIRNA Assays ›
Gene Symbol:
TSLP
Gene Name:
thymic stromal lymphopoietin
Gene Aliases:
-
Chromosome Location:
Chr.5: 111070080 - 111078024 on Build GRCh38
UniGene ID:
Hs.389874
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s40073

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TSLP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033035.4 NP_149024.1 3 541
NM_138551.4 NP_612561.2 1 323
NR_045089.1 4 1746
XM_011543698.1 XP_011542000.1 2 145
GenBank mRNA AF338732.1 - 3 375
AL833421.1 - 1 303
AY037115.1 - 3 541
BC016720.1 - 1 256
BC022894.1 - 1 295
BC040592.1 - 4 1747 5269764
BF245025.1 - 1 310
BU571002.1 - 1 326

Back To Top

More Information


Gene Ontology Categories:

Process(es)

cytokine activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-gm6kg:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0