siRNA Details
| Gene Symbol: THAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195752.1 | NP_001182681.1 | 2 | 279 | ||
| NM_001195753.1 | NP_001182682.1 | 3 | 369 | |||
| NM_138350.3 | NP_612359.2 | 2 | 279 | |||
| XM_005263532.3 | XP_005263589.1 | 3 | 589 | |||
| XM_011542400.2 | XP_011540702.1 | 1 | 78 | |||
| XM_011542401.2 | XP_011540703.1 | 2 | 237 | |||
| XM_017002761.1 | XP_016858250.1 | 2 | 237 | |||
| GenBank mRNA | AK096217.1 | - | 2 | 237 | ||
| BC092427.1 | - | 3 | 369 | 30376814 | ||
| BE905510.1 | - | 339 | ||||

THAP3
Genomic Map