siRNA Details
| Gene Symbol: ZNF799 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080821.2 | NP_001074290.1 | 4 | 1009 | ||
| NM_001322497.1 | NP_001309426.1 | 4 | 1459 | |||
| NM_001322498.1 | NP_001309427.1 | 5 | 1127 | |||
| XM_017027452.1 | XP_016882941.1 | 3 | 802 | |||
| GenBank mRNA | AK027660.1 | - | 4 | 1459 | ||
| AL832890.1 | - | 3 | 2499 | |||

ZNF799
Genomic Map