This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DPP9 SIRNA Assays ›
Gene Symbol:
DPP9
Gene Name:
dipeptidyl peptidase 9
Gene Aliases:
DP9, DPLP9, DPRP-2, DPRP2
Chromosome Location:
Chr.19: 4675226 - 4723843 on Build GRCh38
UniGene ID:
Hs.515081
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s40557

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DPP9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139159.4 NP_631898.3 13 1726
XM_005259673.2 XP_005259730.1 14 1835
XM_011528405.1 XP_011526707.1 15 1887
XM_011528406.1 XP_011526708.1 16 1997
XM_011528408.1 XP_011526710.1 13 1672
XM_011528410.1 XP_011526712.1 14 1887
XM_017027453.1 XP_016882942.1 15 2477
XM_017027454.1 XP_016882943.1 16 2587
GenBank mRNA AF452102.2 - 13 1746
AF542510.2 - 13 1705
AK054656.1 - 2 266
AK122645.1 - 7 776
AK122654.1 - 13 1729
AK131100.1 - 10 1295
AK131499.1 - 12 1239
AY172660.1 - 10 1391
AY374518.1 - 10 1381
BC000970.1 - 1 109
BC037948.2 - 13 1663 4577856
CR627380.1 - 5 501
DQ417928.1 - 11 1468

Back To Top

More Information


Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
aminopeptidase activity
serine-type peptidase activity
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline