siRNA Details
| Gene Symbol: ZNF830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052857.3 | NP_443089.3 | 1 | 103 | ||
| GenBank mRNA | AK055707.1 | - | 1 | 87 | ||
| AK315671.1 | - | 1 | 103 | |||
| BC002913.2 | - | 1 | 48 | |||
| BC011584.1 | - | 1 | 48 | |||
| BG766126.1 | - | 75 | ||||
| DB030670.1 | - | 1 | 103 | |||

ZNF830
Genomic Map