This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SLC22A9 SIRNA Assays ›
Gene Symbol:
SLC22A9
Gene Name:
solute carrier family 22 member 9
Gene Aliases:
HOAT4, OAT4, OAT7, UST3H, ust3
Chromosome Location:
Chr.11: 63368871 - 63410878 on Build GRCh38
UniGene ID:
Hs.502772
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s41562

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SLC22A9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080866.2 NP_543142.2 3 905
XM_017017159.1 XP_016872648.1 3 1823
GenBank mRNA AB062418.1 - 3 738
AB074812.1 - 3 886
AF440402.1 - 3 873
AJ295270.1 - 3 697
AK074246.1 - 3 905
BC126288.1 - 3 787 8992004

Back To Top

More Information


Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

hormone transport
inorganic anion transport
short-chain fatty acid import
sodium-independent organic anion transport
anion transmembrane transport
inorganic anion exchanger activity
anion:anion antiporter activity
sodium-independent organic anion transmembrane transporter activity
short-chain fatty acid uptake transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-jq955:80/100.66.72.86:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0