This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ESCO1 SIRNA Assays ›
Gene Symbol:
ESCO1
Gene Name:
establishment of sister chromatid cohesion N-acetyltransferase 1
Gene Aliases:
A930014I12Rik, CTF, ECO1, EFO1, ESO1
Chromosome Location:
Chr.18: 21529281 - 21601015 on Build GRCh38
UniGene ID:
Hs.464733
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s41632

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ESCO1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052911.2 NP_443143.2 4 1637
XM_011525798.1 XP_011524100.1 4 1767
XM_011525799.2 XP_011524101.1 4 1900
GenBank mRNA AB067498.1 - 4 1573
AK096746.1 - 4 1639
AL832041.1 - 4 1789
AL834200.1 - 1 345
BC089426.1 - 1 1057 3090307

Back To Top

More Information


Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

regulation of DNA replication
sister chromatid cohesion
post-translational protein acetylation
N-acetyltransferase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline