This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM200A SIRNA Assays ›
Gene Symbol:
TMEM200A
Gene Name:
transmembrane protein 200A
Gene Aliases:
KIAA1913, TTMA, TTMC
Chromosome Location:
Chr.6: 130366281 - 130443424 on Build GRCh38
UniGene ID:
Hs.591341
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s41637

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM200A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258276.1 NP_001245205.1 2 1179
NM_001258277.1 NP_001245206.1 3 1243
NM_001258278.1 NP_001245207.1 2 1198
NM_052913.2 NP_443145.1 2 3297
XM_005266816.2 XP_005266873.1 4 1322
XM_011535419.2 XP_011533721.1 2 999
XM_017010230.1 XP_016865719.1 2 1974
GenBank mRNA AB052099.1 - 2 3297
AB067500.1 - 3 1790
AK290166.1 - 2 1192
AK294359.1 - 1 622
AK315824.1 - 2 1075
BC044246.1 - 3 1062 5760073

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-8p9zv:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline