This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DMPK SIRNA Assays ›
Gene Symbol:
DMPK
Gene Name:
dystrophia myotonica protein kinase
Gene Aliases:
DM, DM1, DM1PK, DMK, MDPK, MT-PK
Chromosome Location:
Chr.19: 45769709 - 45782557 on Build GRCh38
UniGene ID:
Hs.631596
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s4166

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DMPK
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081560.2 NP_001075029.1 7, 8 1079
NM_001081562.2 NP_001075031.1 7, 8 1079
NM_001081563.2 NP_001075032.1 6, 7 1448
NM_001288764.1 NP_001275693.1 8, 9 1138
NM_001288765.1 NP_001275694.1 5, 6 719
NM_001288766.1 NP_001275695.1 7, 8 1079
NM_004409.4 NP_004400.4 7, 8 1079
GenBank mRNA AB209363.1 - 7, 8 1922
AK297299.1 - 5, 6 670
AK302874.1 - 7, 8 1012
BC026328.1 - 8, 9 1133 4792104
BC062553.1 - 7, 8 1000 6480902
KJ534826.1 - 6, 7 904
KJ534827.1 - 6, 7 904
KJ534828.1 - 6, 7 904
L19268.1 - 7, 8 1001
M87312.1 - 6, 7 1449
M94203.1 - 7, 8 751
S72883.1 - 7, 8 910

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of sodium ion transport
protein phosphorylation
cellular calcium ion homeostasis
nuclear envelope organization
regulation of heart contraction
muscle cell apoptotic process
regulation of myotube differentiation
regulation of skeletal muscle contraction by calcium ion signaling
regulation of excitatory postsynaptic membrane potential involved in skeletal muscle contraction
peptidyl-serine phosphorylation
regulation of catalytic activity
regulation of synapse structural plasticity
regulation of cardiac conduction
protein serine/threonine kinase activity
protein binding
ATP binding
myosin phosphatase regulator activity
heat shock protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-mhfk8:80/100.66.78.154:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0