This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZG16B SIRNA Assays ›
Gene Symbol:
ZG16B
Gene Name:
zymogen granule protein 16B
Gene Aliases:
EECP, HRPE773, JCLN2, PAUF, PRO1567
Chromosome Location:
Chr.16: 2830172 - 2832284 on Build GRCh38
UniGene ID:
Hs.105887
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s42712

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZG16B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145252.2 NP_660295.2 3 285
GenBank mRNA AK311810.1 - 3 153
AY359021.1 - 3 227
BC009722.1 - 3 225 3879368
BQ924772.1 - 3 288
EF067313.1 - 3 184
JN411265.1 - 3 226

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

retina homeostasis
molecular_function
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-5djdz:80/100.66.79.206:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline