This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HMGB4 SIRNA Assays ›
Gene Symbol:
HMGB4
Gene Name:
high mobility group box 4
Gene Aliases:
dJ1007G16.5
Chromosome Location:
Chr.1: 33860475 - 33864791 on Build GRCh38
UniGene ID:
Hs.568628
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s43188

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: HMGB4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145205.5 NP_660206.2 2 2165
NR_033264.1 2 450
GenBank mRNA AK097707.1 - 2 2166
AK130859.1 - 1 584
AK310465.1 - 2 450
AK311985.1 - 1 551
BC021180.2 - 1 535 5266418
BC070148.1 - 2 450 6617009
BI827031.1 - 1 537
DQ207367.1 - 1 535

Back To Top

More Information


Gene Ontology Categories:

Process(es)

DNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-cdktp:80/100.66.78.54:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline