This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF474 SIRNA Assays ›
Gene Symbol:
ZNF474 LOC100505841
Gene Name:
zinc finger protein 474
zinc finger protein 474-like
Gene Aliases:
-
-
Chromosome Location:
Chr.5: 122129513 - 122153571 on Build GRCh38
Chromosome Location:
Chr.5: 122129546 - 122182663 on Build GRCh38
UniGene ID:
Hs.729673
Hs.733368
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID s43816

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF474
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207317.2 NP_997200.1 2 609
GenBank mRNA AK057483.1 - 2 603
AK290985.1 - 2 539
AY461732.1 - 2 602
BC133022.1 - 1 284 40147044
BC133026.1 - 1 284 40147048
Gene Symbol: LOC100505841
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017008912.1 XP_016864401.1 2 576

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Process(es)

metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline