This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other WDR36 SIRNA Assays ›
Gene Symbol:
WDR36
Gene Name:
WD repeat domain 36
Gene Aliases:
GLC1G, TA-WDRP, TAWDRP, UTP21
Chromosome Location:
Chr.5: 111092172 - 111130502 on Build GRCh38
UniGene ID:
Hs.533237
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s43866

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 388.00
Your Price
Online offer: 287.12
287.12
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: WDR36
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139281.2 NP_644810.1 7 898
GenBank mRNA AF385437.1 - 7 898
AL832494.1 - 6 704
BC133025.1 - 7 858 40147047
BC136517.1 - 7 858 9020507
CR749211.1 - 2 98

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

retina homeostasis
rRNA processing
visual perception
biological_process
regulation of axon extension
response to stimulus
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xg9l8:80/100.66.77.189:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline