This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OR13C5 SIRNA Assays ›
Gene Symbol:
OR13C5 OR13C2
Gene Name:
olfactory receptor family 13 subfamily C member 5
olfactory receptor family 13 subfamily C member 2
Gene Aliases:
OR9-11
OR37K
Chromosome Location:
Chr.9: 104598457 - 104599413 on Build GRCh38
Chromosome Location:
Chr.9: 104604671 - 104605627 on Build GRCh38
UniGene ID:
Hs.553598
Hs.553789
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID s44134

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OR13C5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004482.1 NP_001004482.1 1 406
GenBank mRNA BC136842.1 - 1 500 9020833
BC136843.1 - 1 500 9020834
Gene Symbol: OR13C2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004481.1 NP_001004481.1 1 406
GenBank mRNA BC136824.1 - 1 449 9020815
BC136826.1 - 1 449 9020817

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-gm6kg:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0