This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RNF113B SIRNA Assays ›
Gene Symbol:
RNF113B
Gene Name:
ring finger protein 113B
Gene Aliases:
RNF161, ZNF183L1, bA10G5.1
Chromosome Location:
Chr.13: 98175785 - 98177267 on Build GRCh38
UniGene ID:
Hs.296045
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s44264

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF113B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178861.4 NP_849192.1 1 707
GenBank mRNA AF539427.1 - 1 688
BC017585.2 - 1 707 4828542
BC025388.1 - 1 700 4824788

Back To Top

More Information


Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

snoRNA splicing
mRNA cis splicing, via spliceosome
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline