siRNA Details
| Gene Symbol: RNF113B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178861.4 | NP_849192.1 | 1 | 746 | ||
| GenBank mRNA | AF539427.1 | - | 1 | 727 | ||
| BC017585.2 | - | 1 | 746 | 4828542 | ||
| BC025388.1 | - | 1 | 739 | 4824788 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF113B| Gene Symbol: RNF113B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178861.4 | NP_849192.1 | 1 | 746 | ||
| GenBank mRNA | AF539427.1 | - | 1 | 727 | ||
| BC017585.2 | - | 1 | 746 | 4828542 | ||
| BC025388.1 | - | 1 | 739 | 4824788 | ||