This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LYRM5 SIRNA Assays ›
Gene Symbol:
LYRM5
Gene Name:
LYR motif containing 5
Gene Aliases:
-
Chromosome Location:
Chr.12: 25195216 - 25205051 on Build GRCh38
UniGene ID:
Hs.209151
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s44641

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LYRM5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001660.2 NP_001001660.2 3 369
XM_005253319.4 XP_005253376.1 3 372
XM_005253320.4 XP_005253377.1 4 476
XM_017018850.1 XP_016874339.1 3 1790
GenBank mRNA AK057730.1 - 3 373
AK095164.1 - 2 1870
BC071769.1 - 3 303 4246777
BC100276.1 - 3 320 6178563
BG771497.1 - 3 375

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-gm6kg:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0