This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ZNF420 SIRNA Assays ›
Gene Symbol:
ZNF420
Gene Name:
zinc finger protein 420
Gene Aliases:
APAK
Chromosome Location:
Chr.19: 37007815 - 37130322 on Build GRCh38
UniGene ID:
Hs.444992
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s45081

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF420
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006723038.3 XP_006723101.1 5 652
XM_006723039.3 XP_006723102.1 4 702
XM_011526503.2 XP_011524805.1 5 762
XM_011526504.2 XP_011524806.1 4 576
XM_011526505.2 XP_011524807.1 6 734
XM_011526507.2 XP_011524809.1 3 410
XM_011526508.2 XP_011524810.1 2 368
XM_011526510.2 XP_011524812.1 3 547
XM_011526511.2 XP_011524813.1 5 652
XM_017026336.1 XP_016881825.1 4 576
XM_017026337.1 XP_016881826.1 5 667
XM_017026338.1 XP_016881827.1 2 322
XM_017026339.1 XP_016881828.1 6 734
XM_017026340.1 XP_016881829.1 6 734
GenBank mRNA AK056753.1 - 5 631
AK315728.1 - 5 647
BC026210.2 - 5 586 3028503

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of apoptotic process
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-stzfq:80/100.66.77.128:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0