This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LOC101929023 SIRNA Assays ›
Gene Symbol:
LOC101929023
Gene Name:
uncharacterized LOC101929023
Gene Aliases:
-
Chromosome Location:
Chr.1: 116423724 - 116478826 on Build GRCh38
UniGene ID:
Hs.668091
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s45210

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LOC101929023
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_125972.1 1, 2 52
GenBank mRNA BX097658.1 - 1, 2 35

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-5p4xw:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline