This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other JAKMIP3 SIRNA Assays ›
Gene Symbol:
JAKMIP3
Gene Name:
Janus kinase and microtubule interacting protein 3
Gene Aliases:
C10orf14, C10orf39, Jamip3, NECC2, bA140A10.5
Chromosome Location:
Chr.10: 132036068 - 132184852 on Build GRCh38
UniGene ID:
Hs.106254
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s458238

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: JAKMIP3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105521.3 NP_001098991.1 21 2689
NM_001323086.1 NP_001310015.1 21 2695
NM_001323087.1 NP_001310016.1 21 2695
NM_001323088.1 NP_001310017.1 21 2689
NM_001323089.1 NP_001310018.1 20 2467
NM_001323090.1 NP_001310019.1 21 2689
XM_005252674.4 XP_005252731.1 21 4683
XM_005252676.4 XP_005252733.1 20 4467
XM_011539676.2 XP_011537978.1 21 2941
XM_011539682.2 XP_011537984.1 20 4473
XM_017016095.1 XP_016871584.1 21 4689
XM_017016096.1 XP_016871585.1 21 2934
XM_017016097.1 XP_016871586.1 20 4461
GenBank mRNA AK126964.1 - 11 976
AL832756.1 - 11 1837

Back To Top

More Information


Gene Ontology Categories:

Process(es)

microtubule binding
kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-4spjb:80/100.66.76.244:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0