siRNA Details
| Gene Symbol: WBSCR27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152559.2 | NP_689772.2 | 6 | 905 | ||
| XM_017011777.1 | XP_016867266.1 | 6 | 985 | |||
| XM_017011778.1 | XP_016867267.1 | 6 | 1107 | |||
| GenBank mRNA | AF534110.1 | - | 6 | 916 | ||
| AY354928.1 | - | 6 | 915 | |||
| BC030295.2 | - | 5 | 869 | 5396423 | ||

WBSCR27
Genomic Map