This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LINGO2 SIRNA Assays ›
Gene Symbol:
LINGO2
Gene Name:
leucine rich repeat and Ig domain containing 2
Gene Aliases:
LERN3, LRRN6C
Chromosome Location:
Chr.9: 27937617 - 29213981 on Build GRCh38
UniGene ID:
Hs.444665
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s46014

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LINGO2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258282.1 NP_001245211.1 7 1205
NM_152570.2 NP_689783.1 7 1285
XM_011517719.1 XP_011516021.1 6 2153
XM_011517724.2 XP_011516026.1 5 5172
XM_011517728.2 XP_011516030.1 2 4951
XM_017014303.1 XP_016869792.1 11 5056
XM_017014304.1 XP_016869793.1 4 7576
XM_017014305.1 XP_016869794.1 5 7159
XM_017014306.1 XP_016869795.1 3 4320
XM_017014307.1 XP_016869796.1 2 1793
GenBank mRNA AK056372.1 - 7 1285
AK172834.1 - 1 285
AK290972.1 - 2 816
AY358114.1 - 2 839
BC137514.1 - 7 1145 9021514
BC144676.1 - 7 1145 9053210

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107926.
This siRNA also targets the following transcripts in Mouse: NM_001166001, NM_001166000, NM_001165999, NM_175516, BC137866, and others.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

axonogenesis
positive regulation of synapse assembly

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-v4fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline