This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other USP51 SIRNA Assays ›
Gene Symbol:
USP51
Gene Name:
ubiquitin specific peptidase 51
Gene Aliases:
-
Chromosome Location:
Chr.X: 55484524 - 55489875 on Build GRCh38
UniGene ID:
Hs.40061
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s46136
Validated

Purification
Size
Availability Inventoried
Catalog # 4390824
Price 7,898.00
Your Price
Online offer:
7898.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: USP51
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201286.3 NP_958443.1 2 903
XM_017029299.1 XP_016884788.1 3 1170
XM_017029300.1 XP_016884789.1 3 1053
XM_017029301.1 XP_016884790.1 2 973
GenBank mRNA AJ583823.2 - 1 824
BC035907.1 - 2 914

Back To Top

More Information


Matching Taqman Gene Expression Assay:


Hs01651394_s1

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
histone deubiquitination
protein deubiquitination
thiol-dependent ubiquitin-specific protease activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0