This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other LDHAL6A SIRNA Assays ›
Gene Symbol:
LDHAL6A
Gene Name:
lactate dehydrogenase A like 6A
Gene Aliases:
LDH6A
Chromosome Location:
Chr.11: 18455827 - 18479600 on Build GRCh38
UniGene ID:
Hs.668877
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s46184

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LDHAL6A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144071.1 NP_001137543.1 3 422
NM_144972.4 NP_659409.2 2 1015
XM_005252805.4 XP_005252862.2 3 304
XM_011519922.2 XP_011518224.1 3 306
XM_011519923.2 XP_011518225.1 3 393
XM_011519924.2 XP_011518226.1 3 250
XM_011519925.1 XP_011518227.1 3 232
GenBank mRNA AK131523.1 - 2 958
AY581313.1 - 3 422
BC014340.1 - 3 426 3935569

Back To Top

More Information


Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
carboxylic acid metabolic process
oxidation-reduction process
L-lactate dehydrogenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline