siRNA Details
| Gene Symbol: ZNF883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101338.1 | NP_001094808.1 | 5 | 1280 | ||
| GenBank mRNA | AK095843.1 | - | 5 | 1280 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF883| Gene Symbol: ZNF883 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101338.1 | NP_001094808.1 | 5 | 1280 | ||
| GenBank mRNA | AK095843.1 | - | 5 | 1280 | ||