siRNA Details
| Gene Symbol: DCDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020869.3 | NP_065920.2 | 12 | 1801 | ||
| GenBank mRNA | AB040926.1 | - | 2 | 327 | ||
| BX647348.1 | - | 12 | 1801 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DCDC5| Gene Symbol: DCDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020869.3 | NP_065920.2 | 12 | 1801 | ||
| GenBank mRNA | AB040926.1 | - | 2 | 327 | ||
| BX647348.1 | - | 12 | 1801 | |||