This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
See other EPB41 SIRNA Assays ›
Gene Symbol:
EPB41
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
4.1R, EL1, HE
Chromosome Location:
Chr.1: 28887091 - 29120046 on Build GRCh38
UniGene ID:
Hs.175437
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4702
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
388.00
Your Price
Online offer:
287.12
287.12
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166005.1
NP_001159477.1
11
1645
NM_001166006.1
NP_001159478.1
11
1718
NM_001166007.1
NP_001159479.1
11
1628
NM_004437.3
NP_004428.1
12
1708
NM_203342.2
NP_976217.1
12
1725
NM_203343.2
NP_976218.1
10
1540
XM_005245753.1
XP_005245810.1
11
1720
XM_005245757.1
XP_005245814.1
11
1720
XM_005245760.1
XP_005245817.1
11
1720
XM_005245761.1
XP_005245818.1
11
1720
XM_005245763.1
XP_005245820.1
11
1720
XM_005245764.1
XP_005245821.1
11
1720
XM_005245765.1
XP_005245822.1
11
1720
XM_005245768.1
XP_005245825.1
11
1720
XM_005245769.1
XP_005245826.1
11
1720
XM_005245770.1
XP_005245827.1
11
1720
XM_005245772.4
XP_005245829.1
11
1720
XM_005245773.4
XP_005245830.1
11
1720
XM_005245774.1
XP_005245831.1
11
1720
XM_006710434.1
XP_006710497.1
11
1720
XM_006710439.1
XP_006710502.1
11
1720
XM_011540956.1
XP_011539258.1
11
1720
XM_011540957.1
XP_011539259.1
11
1720
XM_011540958.1
XP_011539260.1
11
1720
XM_011540959.1
XP_011539261.1
11
1720
XM_011540960.1
XP_011539262.1
11
1720
XM_011540961.1
XP_011539263.1
11
1720
XM_011540962.1
XP_011539264.1
11
1720
XM_011540963.2
XP_011539265.1
11
1720
XM_011540964.1
XP_011539266.1
11
1616
XM_011540965.2
XP_011539267.1
11
1720
XM_017000581.1
XP_016856070.1
10
1615
XM_017000582.1
XP_016856071.1
10
1615
XM_017000583.1
XP_016856072.1
11
1720
XM_017000584.1
XP_016856073.1
11
2011
XM_017000585.1
XP_016856074.1
11
2008
XM_017000586.1
XP_016856075.1
11
1720
XM_017000587.1
XP_016856076.1
11
1720
XM_017000588.1
XP_016856077.1
11
1720
XM_017000589.1
XP_016856078.1
10
1615
XM_017000590.1
XP_016856079.1
10
1615
XM_017000591.1
XP_016856080.1
11
1720
XM_017000592.1
XP_016856081.1
11
1720
XM_017000593.1
XP_016856082.1
11
1616
XM_017000594.1
XP_016856083.1
11
1616
XM_017000595.1
XP_016856084.1
11
1616
XM_017000596.1
XP_016856085.1
11
1616
XM_017000597.1
XP_016856086.1
11
1616
XM_017000598.1
XP_016856087.1
11
1616
XM_017000599.1
XP_016856088.1
11
1616
XM_017000600.1
XP_016856089.1
11
1616
XM_017000601.1
XP_016856090.1
11
1616
XM_017000602.1
XP_016856091.1
11
1616
XM_017000603.1
XP_016856092.1
11
1616
XM_017000604.1
XP_016856093.1
11
1616
GenBank mRNA
AB209649.1
-
11
1628
AF156225.1
-
10
1518
AK310655.1
-
10
1556
BC039079.1
-
11
1715
5167140
BC096104.1
-
8
934
BC096105.1
-
10
1252
40001729
BC113841.1
-
10
1252
BC114450.1
-
10
1252
J03796.1
-
10
1459
M14993.1
-
12
1689
M61733.1
-
12
1706
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
blood circulation
actin cytoskeleton organization
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural constituent of cytoskeleton
protein binding
calmodulin binding
1-phosphatidylinositol binding
spectrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline