This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ANKRD23 SIRNA Assays ›
Gene Symbol:
ANKRD23
Gene Name:
ankyrin repeat domain 23
Gene Aliases:
DARP, MARP3
Chromosome Location:
Chr.2: 96837914 - 96844021 on Build GRCh38
UniGene ID:
Hs.643430
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s47250

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ANKRD23
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144994.7 NP_659431.5 7, 8 745
GenBank mRNA AF492401.1 - 7, 8 721
AJ315766.1 - 6, 7 689
AK092564.1 - 6, 7 619
AY196212.1 - 7, 8 729
BC107056.2 - 7, 8 740 40005436

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to mechanical stimulus
titin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mlg7h:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline