siRNA Details
| Gene Symbol: C17orf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175734.4 | NP_783861.3 | 3 | 779 | ||
| XM_006721471.3 | XP_006721534.1 | 3 | 514 | |||
| XM_011523722.2 | XP_011522024.1 | 3 | 1367 | |||
| GenBank mRNA | BC031286.1 | - | 3 | 779 | ||
| BC044816.1 | - | 3 | 416 | |||
| BC048010.1 | - | 3 | 779 | |||

C17orf74
Genomic Map