This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SLC16A9 SIRNA Assays ›
Gene Symbol:
SLC16A9
Gene Name:
solute carrier family 16 member 9
Gene Aliases:
C10orf36, MCT9
Chromosome Location:
Chr.10: 59650764 - 59710079 on Build GRCh38
UniGene ID:
Hs.499709
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s47916

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SLC16A9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323977.1 NP_001310906.1 5 565
NM_001323978.1 NP_001310907.1 6 993
NM_001323979.1 NP_001310908.1 5 680
NM_001323980.1 NP_001310909.1 5 910
NM_001323981.1 NP_001310910.1 6 1123
NM_194298.2 NP_919274.1 5 1040
XM_017015883.1 XP_016871372.1 5 1040
XM_017015884.1 XP_016871373.1 6 1373
GenBank mRNA AK125791.1 - 5 700
BC126238.1 - 4 629 8991954
BC130567.1 - 4 629 40146598

Back To Top

More Information


Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

monocarboxylic acid transport
urate metabolic process
transmembrane transport
protein binding
monocarboxylic acid transmembrane transporter activity
symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-xp8mr:80/100.66.76.90:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0