siRNA Details
| Gene Symbol: ANGPTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178127.4 | NP_835228.2 | 2, 3 | 699 | ||
| XM_011542735.2 | XP_011541037.1 | 2, 3 | 693 | |||
| XM_017017466.1 | XP_016872955.1 | 2, 3 | 693 | |||
| GenBank mRNA | AK291523.1 | - | 1, 2 | 163 | ||
| AY169281.1 | - | 1, 2 | 157 | |||
| AY358827.1 | - | 1, 2 | 147 | |||
| BC049170.1 | - | 2, 3 | 702 | 5285447 | ||
| BC053957.1 | - | 2, 3 | 702 | |||

ANGPTL5
Genomic Map