This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PROSER2 SIRNA Assays ›
Gene Symbol:
PROSER2
Gene Name:
proline and serine rich 2
Gene Aliases:
C10orf47
Chromosome Location:
Chr.10: 11823341 - 11872277 on Build GRCh38
UniGene ID:
Hs.435775
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s48540

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PROSER2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153256.3 NP_694988.3 3 360
XM_011519438.2 XP_011517740.1 4 679
XM_017016028.1 XP_016871517.1 3 1346
GenBank mRNA BC017269.2 - 3 360 4588686
BC048800.1 - 3 360 5215506

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline