siRNA Details
| Gene Symbol: EVI2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003927.2 | NP_001003927.1 | 3 | 542 | ||
| NM_014210.3 | NP_055025.2 | 2 | 401 | |||
| GenBank mRNA | AK295311.1 | - | 3 | 477 | ||
| AK312348.1 | - | 2 | 354 | |||
| AV724051.1 | - | 2 | 322 | |||
| BC035572.1 | - | 2 | 289 | 5229926 | ||
| BG699778.1 | - | 3 | 442 | |||
| BG700097.1 | - | 3 | 498 | |||
| BI597493.1 | - | 2 | 359 | |||
| DA364537.1 | - | 2 | 401 | |||

EVI2A
Genomic Map