This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IGSF22 SIRNA Assays ›
Gene Symbol:
IGSF22
Gene Name:
immunoglobulin superfamily member 22
Gene Aliases:
IGFN2
Chromosome Location:
Chr.11: 18704305 - 18726939 on Build GRCh38
UniGene ID:
Hs.434152
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s49171

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IGSF22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173588.3 NP_775859.3 10 1299
XM_011520030.2 XP_011518332.1 8 4203
XM_011520031.2 XP_011518333.1 8 4203
XM_017017607.1 XP_016873096.1 9 4094
XM_017017608.1 XP_016873097.1 9 2117
GenBank mRNA AK095113.1 - 10 1299
BC115396.1 - 10 1250
BC115397.1 - 10 1250 40068579

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-dz54l:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline