siRNA Details
| Gene Symbol: FAM150B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002919.2 | NP_001002919.2 | 4, 5 | 738 | ||
| XM_005264689.4 | XP_005264746.1 | 4, 5 | 751 | |||
| XM_006711879.3 | XP_006711942.1 | 4, 5 | 752 | |||
| GenBank mRNA | AY358517.1 | - | 4, 5 | 207 | ||
| BC033490.1 | - | 4, 5 | 612 | |||

FAM150B
Genomic Map