This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FBXW12 SIRNA Assays ›
Gene Symbol:
FBXW12
Gene Name:
F-box and WD repeat domain containing 12
Gene Aliases:
FBW12, FBXO12, FBXO35
Chromosome Location:
Chr.3: 48372219 - 48394830 on Build GRCh38
UniGene ID:
Hs.288793
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID s49856

Purification
Size
Availability Inventoried
Catalog # 4392420
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FBXW12
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159927.1 NP_001153399.1 4 471
NM_001159929.1 NP_001153401.1 3 270
NM_207102.2 NP_996985.2 4 444
XM_017006224.1 XP_016861713.1 3 457
GenBank mRNA AY247969.1 - 4 444
BC101303.1 - 3 617 40024696
BC101304.1 - 4 415 40024699
BC101305.1 - 3 214 40024702

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-pkm9c:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0