siRNA Details
| Gene Symbol: DNAI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281428.1 | NP_001268357.1 | 4, 5 | 524 | ||
| NM_012144.3 | NP_036276.1 | 4, 5 | 524 | |||
| XM_006716758.3 | XP_006716821.1 | 3, 4 | 222 | |||
| XM_011517846.1 | XP_011516148.1 | 4, 5 | 524 | |||
| XM_011517847.2 | XP_011516149.1 | 4, 5 | 524 | |||
| XM_011517848.1 | XP_011516150.1 | 4, 5 | 524 | |||
| XM_011517849.1 | XP_011516151.1 | 4, 5 | 524 | |||
| XM_011517850.2 | XP_011516152.1 | 4, 5 | 524 | |||
| XM_017014625.1 | XP_016870114.1 | 4, 5 | 524 | |||
| GenBank mRNA | AF091619.1 | - | 4, 5 | 441 | ||
| AK290696.1 | - | 4, 5 | 448 | |||
| AK314639.1 | - | 4, 5 | 436 | |||
| BC030583.2 | - | 4, 5 | 452 | 4837991 | ||
| BM013886.1 | - | 4, 5 | 464 | |||
| DB033871.1 | - | 4, 5 | 448 | |||
| DB046118.1 | - | 4, 5 | 525 | |||

DNAI1
Genomic Map