siRNA Details
| Gene Symbol: AADACL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207365.3 | NP_997248.2 | 5 | 998 | ||
| XM_011512753.1 | XP_011511055.1 | 5 | 1026 | |||
| GenBank mRNA | BC065724.1 | - | 5 | 991 | 6663150 | |
| BX118873.1 | - | 1 | 205 | |||
| BX647585.1 | - | 4 | 775 | |||

AADACL2
Genomic Map