This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other H3F3A SIRNA Assays ›
Gene Symbol:
H3F3A H3F3AP4
Gene Name:
H3 histone, family 3A
H3 histone, family 3A, pseudogene 4
Gene Aliases:
H3.3A, H3F3
p13
Chromosome Location:
Chr.1: 226062707 - 226072002 on Build GRCh38
Chromosome Location:
Chr.2: 174719800 - 174720858 on Build GRCh38
UniGene ID:
Hs.533624
Hs.533624
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID s51241

Purification
Size
Availability Inventoried
Catalog # 4392420
Price 5,366.00
Your Price
Online offer:
5366.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: H3F3A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002107.4 NP_002098.1 3 312
GenBank mRNA AK293541.1 - 3 294
AK311905.1 - 3 312
BC013857.1 - 1 180
BC029405.1 - 3 294 4077662
BC038989.1 - 1 214 6014954
BC081561.1 - 1 285 5162817
BC095447.1 - 3 294 30718542
BM760856.1 - 3 299
M11353.1 - 2 558
M11354.1 - 3 292
Gene Symbol: H3F3AP4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002315.1 1 292
GenBank mRNA BC066901.1 - 1 288
BC067757.1 - 1 294
BC081560.1 - 1 285 3884607

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_002729644, XM_576283, XM_003750348, XM_002728043, BC086580, and others.

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing at rDNA
nucleosome assembly
DNA replication-independent nucleosome assembly
brain development
blood coagulation
response to hormone
positive regulation of cell growth
gene silencing by RNA
telomere organization
cellular protein metabolic process
negative regulation of gene expression, epigenetic
positive regulation of gene expression, epigenetic
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
protein binding
nucleosomal DNA binding
histone binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0